Drum pumps, gas-tight, with flexible discharge
Supplier: BURKLE
Certificates
About this item
Gas tight barrel pump with flexible discharge hose for filling off in to large canisters and containers.
- Convenient transfer as the canister can stay on the floor
- Pump can be hermetically sealed with two ball valves
Delivery information: Includes gas-tight barrel screw joints with 2" fine thread.
Frequently Bought Together