Orders LinkContactUsLinkComponent
Cryostorage boxes, Cryostore™
Cryostorage boxes, Cryostore™
Supplier:   Simport Scientific
Cryostorage boxes, Cryostore™
Supplier:  Simport Scientific
Certificates
Cryostorage boxes, Cryostore™
Cryostorage boxes, Cryostore™
7 Options Available Below
Product Details & Documents
PC, coloured.
  • Writable surfaces
  • Available with four colours
  • Printed with squared numbers
  • Transparent cover allows to see the content of the box
These durable cryogenic storage boxes are designed to be used at temperatures between −196 °C and +121 °C. It can accommodate around 42×T310-10A Cryovials®.
Documents
Product Family Options
Product Information
  • Compartments
    For
    Colour
    W×D×H ext.
    Availability
    Price
  • 5×5
    1 - 2 ml cryotubes
    Blue
    76×76×52 mm
    Log in for availability
    Log in for availability
    1 * 8 ST
    1 * 8 ST - 65,20 €
    65,20 €
    1 * 8 ST
       65,20 €
  • 9×9
    3 - 4 ml cryotubes
    Blue
    133×133×81 mm
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 130,00 €
    130,00 €
    1 * 6 ST
       130,00 €
  • 7×6
    10 ml cryotubes
    Blue
    133×133×95 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 181,00 €
    181,00 €
    1 * 10 ST
       181,00 €
  • 7×6
    10 ml cryotubes
    Green
    133×133×95 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 181,00 €
    181,00 €
    1 * 10 ST
       181,00 €
  • 7×6
    10 ml cryotubes
    Red
    133×133×95 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 181,00 €
    181,00 €
    1 * 10 ST
       181,00 €
  • 7×6
    10 ml cryotubes
    Yellow
    133×133×95 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 181,00 €
    181,00 €
    1 * 10 ST
       181,00 €
  • 9×9
    3 - 5 ml cryotubes
    Blue
    133×133×95 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 108,00 €
    108,00 €
    1 * 5 ST
       108,00 €
  • Discontinued
  • Discontinued