Order Entry
Belgium
ContactUsLinkComponent
Xylene (mixture of isomers) ≥99.5%, CMOS for microelectronic, J.T.Baker®
Flammable
Irritant
Health
Xylene (mixture of isomers) ≥99.5%, CMOS for microelectronic, J.T.Baker®
Catalog # 9493-03
Supplier:  Avantor
CAS Number:  
undefined
Flammable
Irritant
Health
Xylene (mixture of isomers) ≥99.5%, CMOS for microelectronic, J.T.Baker®
Catalog # 9493-03
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    4 L
  • Seq:
    5162

Specifications

Specification Test Results

  • Color (APHA):
    ≤10
  • Residue after Evaporation:
    ≤5 ppm
  • Acidity (µeq/g):
    ≤0.3
  • Water (H₂O):
    ≤0.02 %
  • Chloride (Cl):
    ≤3 ppm
  • Phosphate (PO₄):
    ≤0.5 ppm
  • Trace Impurities - Aluminum (Al):
    ≤50.0 ppb
  • Arsenic and Antimony (as As):
    ≤10.0 ppb
  • Trace Impurities - Barium (Ba):
    ≤20.0 ppb
  • Trace Impurities - Boron (B):
    ≤20.0 ppb
  • Trace Impurities - Cadmium (Cd):
    ≤20.0 ppb
  • Trace Impurities - Calcium (Ca):
    ≤50.0 ppb
  • Trace Impurities - Chromium (Cr):
    ≤50 ppb
  • Trace Impurities - Cobalt (Co):
    ≤20 ppb
  • Trace Impurities - Copper (Cu):
    ≤10.0 ppb
  • Trace Impurities - Gallium (Ga):
    ≤50 ppb
  • Trace Impurities - Germanium (Ge):
    ≤100.0 ppb
  • Trace Impurities - Gold (Au):
    ≤50 ppb
  • Heavy Metals (as Pb):
    ≤1000.0 ppb
  • Trace Impurities - Iron (Fe):
    ≤50.0 ppb
  • Trace Impurities - Lead (Pb):
    ≤100 ppb
  • Trace Impurities - Lithium (Li):
    ≤50.0 ppb
  • Trace Impurities - Magnesium (Mg):
    ≤20 ppb
  • Trace Impurities - Manganese (Mn):
    ≤20.0 ppb
  • Trace Impurities - Nickel (Ni):
    ≤10.0 ppb
  • Trace Impurities - Potassium (K):
    ≤100 ppb
  • Trace Impurities - Silicon (Si):
    ≤200.0 ppb
  • Trace Impurities - Silver (Ag):
    ≤50 ppb
  • Trace Impurities - Sodium (Na):
    ≤100.0 ppb
  • Trace Impurities - Strontium (Sr):
    ≤20.0 ppb
  • Trace Impurities - Tin (Sn):
    ≤100 ppb
  • Trace Impurities - Zinc (Zn):
    ≤20.0 ppb
  • Particle Count - 1.0 µm and greater:
    ≤10 par/ml
  • Total of p-, m-, o-xylene, toluene and ethylbenzene (by GC):
    ≥99.5 %

Specification Test Results

About this item

 :
 :106.17 g/mol
Merck Index:13,10136
Boiling Pt:140 °C (1013 hPa)
Melting Pt:–34 °C
Density:0.86…0.88 g/cm³ (20 °C)
Flash Pt:25 °C
EINECS:215-535-7
UN:1307
ADR:3,III
 :GHS02
 :GHS07
 :GHS08
Storage Temperature:Ambient
CAS Number:1330-20-7
Formula:C₆H₄(CH₃)₂
MDL Number:MFCD00077264