Orders LinkContactUsLinkComponent
Screwthread tubes, attachable, DURAN®
Screwthread tubes, attachable, DURAN®
Supplier:   DWK Life Sciences
Screwthread tubes, attachable, DURAN®
Supplier:  DWK Life Sciences
Certificates
Screwthread tubes, attachable, DURAN®
Screwthread tubes, attachable, DURAN®
5 Options Available Below
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Screw thread GL. It is used by glassblowers in the construction of laboratory glassware.
Caution: These tubes are open at the bottom.
Product Family Options
Product Information
  • Thread
    Tube ext.Ø
    Wall thickness
    Availability
    Price
  • GL 14
    12 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 218,00 €
    218,00 €
    1 * 100 ST
       218,00 €
  • GL 18
    16 mm
    1,8 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 128,00 €
    128,00 €
    1 * 50 ST
       128,00 €
  • GL 25
    22 mm
    1,8 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 231,00 €
    231,00 €
    1 * 50 ST
       231,00 €
  • GL 32
    28 mm
    2,0 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 359,00 €
    359,00 €
    1 * 50 ST
       359,00 €
  • GL 45
    40 mm
    2,3 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 202,00 €
    202,00 €
    1 * 10 ST
       202,00 €