ContactUsLinkComponent
Thermometer clamp, with swivelling bosshead
Thermometer clamp, with swivelling bosshead
Catalog # BOCH5621
Supplier:  BOCHEM
undefined
Thermometer clamp, with swivelling bosshead
Catalog # BOCH5621
Supplier:  BOCHEM
Specifications
  • For:
    Thermometers
  • Spanning width:
    6 - 15 mm
  • Pk:
    1
Specifications
About this item
These chromium plated clamps are used for holding thermometers (Ø 6 to 15 mm). Swivelling bosshead (Ø 16,5 mm) can be rotated and locked at any angle (0 to 360°).