Orders LinkContactUsLinkComponent
Pharma zone sampler, Novartos Multi
Pharma zone sampler, Novartos Multi
Supplier:   BURKLE
Pharma zone sampler, Novartos Multi
Supplier:  BURKLE
Certificates
Pharma zone sampler, Novartos Multi
Pharma zone sampler, Novartos Multi
14 Options Available Below
Product Details & Documents
Novartos is the pharmaceutical zone sampler for sampling in accordance with the FDA guidelines for unit dose sampling. With the Novartos pharmaceutical samplers, multiple individual dose samples (unit dose samples) can be taken from one or more sampling points in a single sampling.
  • Sample inserts fit precisely and can easily be replaced
  • Inserts are available in volumes from 0 to 2,0 ml
  • Insertion depths can easily be read from the laser-engraved depth scale
  • Stainless steel AISI 316L (1.4404) / AISI 316Ti (1.4571)
  • Groove-free and hand-polished
  • Easy cleaning
  • 25 mm Ø
Novartos Multi double sampling:
With Novartos Multi double sampling, a double multi-point sample can be taken from multiple predefined sample zones. Depending on the length, four, seven, or nine sample chambers are each fitted with two sample inserts for the double sampling.
Novartos Multi triple sampling:
With Novartos Multi triple sampling, a triple multi-point sample can be taken from multiple predefined sample zones. Depending on the length, four, seven, or nine sample chambers are each fitted with three sample inserts for the triple sampling.
Ordering information: Please order sample inserts separately.
Documents
Product Family Options
Product Information
  • Length
    No. of chambers
    Ø
    Availability
    Price
  • 129-1782
    Novartos Multi, V4A, double sampling, without inserts, 4 chambers (8 inserts)
    Novartos Multi, V4A, double sampling, without inserts, 4 chambers (8 inserts)
    600 mm
    4
    25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 970,00 €
    2 970,00 €
    1 * 1 ST
       2 970,00 €
  • 300-0126
    Novartos Multi, V4A, double sampling, without inserts, 7 chambers (14 inserts)
    Novartos Multi, V4A, double sampling, without inserts, 7 chambers (14 inserts)
    1000 mm
    7
    25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 580,00 €
    3 580,00 €
    1 * 1 ST
       3 580,00 €
  • 129-1783
    Novartos Multi, V4A, double sampling, without inserts, 9 chambers (18 inserts)
    Novartos Multi, V4A, double sampling, without inserts, 9 chambers (18 inserts)
    1400 mm
    9
    25 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 3 920,00 €
    3 920,00 €
    1 * 1 Pce
       3 920,00 €
  • 129-1779
    Novartos Multi, V4A, triple sampling, without inserts, 4 chambers (12 inserts)
    Novartos Multi, V4A, triple sampling, without inserts, 4 chambers (12 inserts)
    600 mm
    4
    25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 370,00 €
    3 370,00 €
    1 * 1 ST
       3 370,00 €
  • 129-1780
    Novartos Multi, V4A, triple sampling, without inserts, 7 chambers (21 inserts)
    Novartos Multi, V4A, triple sampling, without inserts, 7 chambers (21 inserts)
    1000 mm
    7
    25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4 000,00 €
    4 000,00 €
    1 * 1 ST
       4 000,00 €
  • 129-1781
    Novartos Multi, V4A, triple sampling, without inserts, 9 chambers (27 inserts)
    Novartos Multi, V4A, triple sampling, without inserts, 9 chambers (27 inserts)
    1400 mm
    9
    25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4 460,00 €
    4 460,00 €
    1 * 1 ST
       4 460,00 €
Inserts for Novartos Multi
Product Information
  • Length
    No. of chambers
    Ø
    Availability
    Price
  • 300-0187
    Sampling insert, 0 ml (dummy insert)
    Sampling insert, 0 ml (dummy insert)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 184,00 €
    184,00 €
    1 * 1 ST
       184,00 €
  • BURK5358-0003
    Sampling insert, 0,3 ml
    Sampling insert, 0,3 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 269,00 €
    269,00 €
    1 * 1 ST
       269,00 €
  • 300-0188
    Sampling insert, 0,5 ml
    Sampling insert, 0,5 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 213,00 €
    213,00 €
    1 * 1 ST
       213,00 €
  • BURK5358-0008
    Sampling insert, 0,8 ml
    Sampling insert, 0,8 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 269,00 €
    269,00 €
    1 * 1 ST
       269,00 €
  • 300-0189
    Sampling insert, 1,0 ml
    Sampling insert, 1,0 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 213,00 €
    213,00 €
    1 * 1 ST
       213,00 €
  • 300-0190
    Sampling insert, 1,5 ml
    Sampling insert, 1,5 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 213,00 €
    213,00 €
    1 * 1 ST
       213,00 €
  • BURK5358-0017
    Sampling insert, 1,7 ml
    Sampling insert, 1,7 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 269,00 €
    269,00 €
    1 * 1 ST
       269,00 €
  • 300-0191
    Sampling insert, 2,0 ml
    Sampling insert, 2,0 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 213,00 €
    213,00 €
    1 * 1 ST
       213,00 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Transport case
    Transport case
    312-1141
    Log in for availability
    1 * 1 ST - 202,00 €