Order Entry
Belgium
Orders LinkContactUsLinkComponent
Universal scissors, with plastic handle

Universal scissors, with plastic handle

Blunt/blunt, 210 mm
Catalog # BOCH4251
Supplier:   BOCHEM
Universal scissors, with plastic handle
Catalog # BOCH4251
Supplier:  BOCHEM
Universal scissors, with plastic handle
Universal scissors, with plastic handle

Blunt/blunt, 210 mm

Price
37,70 €
Log in for availability
1 * 1 ST
Product Details & Documents

Universal scissors with plastic handle (PP + TPE) and stainless steel blades for industrial applications.

  • Adjustable knob: adjustable to fit any hand comfortability
  • Hanging hole: easy to carry
  • Blades made of 4 mm stainless steel (HCR 50-54)
  • Serrated blade for gripping materials

Suitable for cutting aluminium and plastic board, brass plates (non-hardened), leather and cardboard.

Specifications
  • Type:
    Blunt/blunt
  • Length:
    210 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.