Order Entry
Belgium
Orders LinkContactUsLinkComponent
Universal scissors, with plastic handle

Universal scissors, with plastic handle

Sharp/blunt, 180 mm
Catalog # BOCH4151
Supplier:   BOCHEM
Universal scissors, with plastic handle
Catalog # BOCH4151
Supplier:  BOCHEM
Universal scissors, with plastic handle
Universal scissors, with plastic handle

Sharp/blunt, 180 mm

Price
9,90 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel bandage scissors with plastic handle.

  • Temperature resistant up to max. 145 °C
  • Sharp-round
Specifications
  • Type:
    Sharp/blunt
  • Length:
    180 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.