
Potassium iodide ≥99.999% (metals basis), TraceSELECT™, Fluka™
30533-100G
:
- Pack type:Plastic bottle
- Pk:100 g
- Seq:3401
- Aluminium (Al):Max. 0.02 ppm
- Arsenic (As):Max. 0.01 ppm
- Barium (Ba):Max. 0.5 ppm
- Calcium (Ca):Max. 0.1 ppm
- Cadmium (Cd):Max. 0.01 ppm
- Cerium (Ce):Max. 0.01 ppm
- Cobalt (Co):Max. 0.01 ppm
- Chromium (Cr):Max. 0.01 ppm
- Copper (Cu):Max. 0.01 ppm
- Europium (Eu):Max. 0.01 ppm
- Iron (Fe):Max. 0.05 ppm
- Mercury (Hg):Max. 5 ppb
- Iridium (Ir):Max. 0.5 ppm
- Lanthanum (La):Max. 0.01 ppm
- Lithium (Li):Max. 0.5 ppm
- Magnesium (Mg):Max. 0.05 ppm
- Manganese (Mn):Max. 0.01 ppm
- Sodium (Na):Max. 5 ppm
- Nickel (Ni):Max. 0.01 ppm
- Lead (Pb):Max. 0.01 ppm
- Palladium (Pd):Max. 0.5 ppm
- Platinum (Pt):Max. 0.5 ppm
- Rhodium (Rh):Max. 0.5 ppm
- Ruthenium (Ru):Max. 0.5 ppm
- Antimony (Sb):Max. 0.01 ppm
- Scandium (Sc):Max. 0.01 ppm
- Samarium (Sm):Max. 0.01 ppm
- Strontium (Sr):Max. 0.1 ppm
- Vanadium (V):Max. 0.1 ppm
- Yttrium (Y):Max. 0.01 ppm
- Ytterbium (Yb):Max. 0.01 ppm
- Zinc (Zn):Max. 0.01 ppm
- Bromide, chloride (as Cl):Max. 50 ppm
- Phosphate (PO₄):Max. 10 ppm
- Total S (as SO₄):Max. 10 ppm
- Total N:Max. 10 ppm
- Iodate (IO₃):Max. 2 ppm
:
:166 g/mol
:13,07727
:1330 °C (1013 hPa)
:686 °C
:3,13 g/cm³ (20 °C)
:Room Temperature
:231-659-4
:GHS08
:7681-11-0
:Kl
:MFCD00011405