Order Entry
Belgium
ContactUsLinkComponent
1-Aminocyclopropanecarboxylic acid, Calbiochem®
1-Aminocyclopropanecarboxylic acid, Calbiochem®
Catalog # 149101-1
CAS Number:  
1-Aminocyclopropanecarboxylic acid, Calbiochem®
Catalog # 149101-1
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :101.11 g/mol
 :M-Clarity™ quality level = MQ100
CAS Number:22059-21-8
Formula:C₄H₇NO₂
MDL Number:MFCD00009944