Creatinine ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98% CAS No.: 60-27-5 Molecular Formula: C4H7N3O Formula Weight: 113.12 Odour: Odourless pH: 7.5-8.5 Melting Point: ca 255° dec. Beilstein Reference: 112061 Merck Reference: 14,2569 MDL No.: MFCD00059730
Synonyms: 2-Amino-1-methyl-1,5-dihydro-4H-imidazol-4-one
- CAS Number:60-27-5
- Formula:C₄H₇N₃O
- MDL Number:MFCD00059730
- Molecular Weight:113.12 g/mol
- Merck Index:14,2569
- Melting Pt:∼ 255 °C (dec.)
- Storage Temperature:Ambient
- EINECS:200-466-7
3 Options Available Below
Product Details & Documents
Purity: 98% CAS No.: 60-27-5 Molecular Formula: C4H7N3O Formula Weight: 113.12 Odour: Odourless pH: 7.5-8.5 Melting Point: ca 255° dec. Beilstein Reference: 112061 Merck Reference: 14,2569 MDL No.: MFCD00059730
Synonyms: 2-Amino-1-methyl-1,5-dihydro-4H-imidazol-4-one
- CAS Number:60-27-5
- Formula:C₄H₇N₃O
- MDL Number:MFCD00059730
- Molecular Weight:113.12 g/mol
- Merck Index:14,2569
- Melting Pt:∼ 255 °C (dec.)
- Storage Temperature:Ambient
- EINECS:200-466-7
Documents
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



