Order Entry
Belgium
Orders LinkContactUsLinkComponent
Creatinine

Creatinine

1 KIT
Catalog # DIAS117119910021
Creatinine
Catalog # DIAS117119910021
CAS Number:  60-27-5
Creatinine

1 KIT

CAS Number:  60-27-5
Price
Demander un prix
Connexion Pour la disponibilité
1 * 1 KIT
Product Details & Documents
Synonymes:  2-Amino-1-methyl-1,5-dihydro-4H-imidazol-4-one
  • Numéro CAS:60-27-5
  • Formule:C₄H₇N₃O
  • Numéro MDL:MFCD00059730
  • Poids moléculaire:113.12 g/mol
  • EINECS:200-466-7
Spécifications

Spécifications

  • Pk:
    1 KIT

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.