Order Entry
Belgium
ContactUsLinkComponent
 
 
 
 
Phenol ≥99% contains no preservatives, white fused crystals, Ultrapure Bioreagent for extraction of nucleic acids, J.T.Baker®
 :  Avantor
 
undefined
 
 
 
Phenol ≥99% contains no preservatives, white fused crystals, Ultrapure Bioreagent for extraction of nucleic acids, J.T.Baker®
 :  Avantor

 

  • For Laboratory,Research,or Manufacturing Use:
    = 99 %
  • Assay (C₆H₅OH):
    ≥ 99 %
  • Melting Point:
    39 - 41 °C
  • Appearance:
    Passes Test
  • DNase Activity (None Detected):
    Passes Test
  • RNase Activity:
    Passes Test
  • pH (saturated):
    4.5 - 6.0
  • Heavy Metals (as Pb):
    ≤ 0.001 %
  • Water (H₂O)(by Karl Fischer titrn):
    ≤ 0.4 %

 

Frequently Bought Together