Order Entry
Belgium
ContactUsLinkComponent
Giemsa's stain solution
Irritant
Giemsa's stain solution
Catalog # CTEH61025/1
Supplier:  CLIN TECH
CAS Number:  
Irritant
Giemsa's stain solution
Catalog # CTEH61025/1
Supplier:  CLIN TECH
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
EINECS:257-438-2
 :GHS07
Storage Temperature:Ambient
CAS Number:51811-82-6
MDL Number:MFCD00081642