Order Entry
Belgium
ContactUsLinkComponent
 
 
 
Pyronin B ≥30% (dye content), Sigma-Aldrich®
  P6892-25G
 :  
undefined
 
 
Pyronin B ≥30% (dye content), Sigma-Aldrich®
  P6892-25G
 :  

 

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance (Colour):
    Dark Green
  • Appearance (Form):
    Powder
  • Infrared spectrum :
    Conforms to Structure
  • Solubility (Colour)
    Dark Red to Purple; c = 1mg/ml, H2O:
    Conforms to Requirements
  • Dye Content:
    ≥ 30.0 %

 

 

 :
 :1042.22 g/mol
 :12,08189
 :174…176 °C
 :M-Clarity™ quality level = MQ200
 :218-429-9
 :GHS08
 :GHS07
 :Room Temperature
 :2150-48-3
 :C₄₂H₅₄N₄O₂·2FeCl₄
 :MFCD03701356