Order Entry
Belgium
ContactUsLinkComponent
 
Sodium acetate
Catalog # RC-089
Supplier:  G-Biosciences
CAS Number:  
Sodium acetate
Catalog # RC-089
Supplier:  G-Biosciences
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :82.03 g/mol
Merck Index:12,08711
Boiling Pt:881.4 °C (1013 hPa)
Melting Pt:324 °C
Density:1.528 g/cm³ (20 °C)
Flash Pt:>250 °C (closed cup)
Storage Temperature:Ambient
EINECS:204-823-8
REACH:01-2119485123-42
CAS Number:127-09-3
Formula:H₃CCOONa
MDL Number:MFCD00012459