Orders LinkContactUsLinkComponent
Vacuum filter holders
Supplier:   Sartorius
Vacuum filter holders
Supplier:  Sartorius
Certificates
2 Options Available Below
Product Details & Documents
These vacuum filter holders feature a glass cylinder, PP base and sealing plug with anodised aluminum closing cap.
  • Vacuum operated
Product Family Options
Product Information
  • Capacity
    For Ø
    Availability
    Price
  • 85 ml
    47 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 117,00 €
    117,00 €
    1 * 1 ST
       117,00 €
  • 130 ml
    47 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 149,00 €
    149,00 €
    1 * 1 ST
       149,00 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Aluminium closing cap
    Aluminium closing cap
    SART6980591
    Log in for availability
    1 * 1 ST - 8,50 €
Product Information
  • Catalog #
    Availability
    Price
  • Closing ring
    Closing ring
    SART6980590
    Log in for availability
    1 * 1 ST - 11,40 €
Product Information
  • Catalog #
    Availability
    Price
  • Glass cylinder, 130 ml
    Glass cylinder, 130 ml
    SART6985060
    Log in for availability
    1 * 100 ST - 77,20 €
Product Information
  • Catalog #
    Availability
    Price
  • Glass cylinder, 85 ml
    Glass cylinder, 85 ml
    SART6980589
    Log in for availability
    1 * 1 ST - 61,80 €
Product Information
  • Catalog #
    Availability
    Price
  • Plug
    Plug
    SART6980588
    Log in for availability
    1 * 10 ST - 39,90 €
Product Information
  • Catalog #
    Availability
    Price
  • PP base
    PP base
    SART6980586
    Log in for availability
    1 * 1 ST - 11,40 €
Product Information
  • Catalog #
    Availability
    Price
  • Flat silicone gasket
    Flat silicone gasket
    SART6980573
    Log in for availability
    1 * 1 ST - 6,50 €
Product Information
  • Catalog #
    Availability
    Price
  • Silicone O-ring
    Silicone O-ring
    SART6980574
    Log in for availability
    1 * 1 ST - 8,50 €