Orders LinkContactUsLinkComponent
Measuring beakers, graduated
Measuring beakers, graduated
Supplier:   BOCHEM
Measuring beakers, graduated
Supplier:  BOCHEM
Certificates
Measuring beakers, graduated
Measuring beakers, graduated
2 Options Available Below
Product Details & Documents
Stainless steel, 18/10, with handle and spout.
  • Good chemical resistance
  • Excellent resistance to heat
  • Smooth surfaces for fast, easy cleaning
Ideal for corrosive materials. Robust and durable.
Documents
Product Family Options
Product Information
  • Capacity
    Height
    Ext.Ø
    Availability
    Price
  • 1000 ml
    140 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 155,00 €
    155,00 €
    1 * 1 ST
       155,00 €
  • 2000 ml
    170 mm
    130 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 314,00 €
    314,00 €
    1 * 1 ST
       314,00 €