Orders LinkContactUsLinkComponent
Dressing jars, with lid
Supplier:   BOCHEM
Dressing jars, with lid
Supplier:  BOCHEM
Certificates
Dressing jars, with lid
Dressing jars, with lid
6 Options Available Below
Product Details & Documents
Jar made of stainless steel, with knob lid.
  • Very good temperature resistance
  • Good chemical resistance
  • Easy to clean
Product Family Options
Product Information
  • Capacity
    Ext.Ø
    Height
    Availability
    Price
  • 100 ml
    60 mm
    60 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 34,90 €
    34,90 €
    1 * 1 ST
       34,90 €
  • 400 ml
    85 mm
    85 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 43,50 €
    43,50 €
    1 * 1 ST
       43,50 €
  • 700 ml
    100 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 57,00 €
    57,00 €
    1 * 1 ST
       57,00 €
  • 1200 ml
    120 mm
    120 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 64,40 €
    64,40 €
    1 * 1 ST
       64,40 €
  • 2500 ml
    150 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 97,00 €
    97,00 €
    1 * 1 ST
       97,00 €
  • 6000 ml
    200 mm
    200 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 156,00 €
    156,00 €
    1 * 1 ST
       156,00 €