Order Entry
Belgium
ContactUsLinkComponent
 
Portable multiparameter colorimeter, Orion™ AQUAfast™ AQ4000
Catalog # 705-0731
Supplier:  Thermo Orion
undefined
Portable multiparameter colorimeter, Orion™ AQUAfast™ AQ4000
Catalog # 705-0731
Supplier:  Thermo Orion

Specifications

  • Light source:
    LED
  • Measurement range:
    0 to 2 A
  • Resolution:
    ±2 nm
  • Description:
    AQUAfast™ AQ4000 colorimeter
  • Model:
    AQ4000
  • Battery life:
    2500 hours (Alkaline); 10000 hours (Lithium)
  • Operating humidity:
    90% at 50 °C max.
  • Cuvettes:
    Ø 24 and 13 mm vials; 16 mm tubes
  • Display:
    Custom LCD
  • Interfaces:
    RS232
  • Keypad:
    12 keys
  • Operating temperature:
    0 to 45 °C
  • Photometric accuracy:
    ±0,005 A at 1,0 A
  • Power supply:
    4×AA batteries
  • Storage capacity:
    Up to 100 datalog points, time and date
  • Wavelengths:
    420; 520; 580; 610 nm
  • Pk:
    1

Specifications

About this item

Measure common colorimetric water and wastewater parameters using the Orion™ AQUAfast™ AQ4000 colorimeter with 65 preprogrammed methods, automatic selection of four wavelengths and timer.

  • Identifies the species to be measured
  • Ensures error-free readings
  • Selects the programme and initiates the measurement
  • Guides user through the setup and measurement with simple operation
  • User can customise calibration and create completely custom methods
  • Up to 12 data points can be used to define custom method
  • Store a maximum of 10 methods in the colorimeter memory

Auto-Test ampoules with Auto-ID barcodes can be used to simplify sample preparation and tests. Ampoules contain pre-measured liquid reagent and fill with the exact volume of sample, colorimeter reads the barcode and automatically identifies the parameter and initiates the test.