ContactUsLinkComponent
Spectroquant reference standard for Picco, Move and Multy colorimeters, Supelco®
Spectroquant reference standard for Picco, Move and Multy colorimeters, Supelco®
Catalog # 1.19301.0001
Supplier:  Merck
CAS Number:  
undefined
Spectroquant reference standard for Picco, Move and Multy colorimeters, Supelco®
Catalog # 1.19301.0001
Supplier:  Merck
CAS Number:  
Specifications
  • Pk:
    1
  • Description:
    Spectroquant® reference standard for chlorine, chlorine dioxide and ozone
Specifications
About this item
A reference standard for chlorine, chlorine dioxide and ozone, that is designed to reassure the user about the accuracy and reliability of the results from the Picco and Multy colorimeters.
Delivery: Standard includes four sealed cells including a Zero cell and stable coloured solutions and is supplied in a handy case with quality data, lot number and expiry date.