Order Entry
Belgium
ContactUsLinkComponent
 
All-layer scratcher
Catalog # 300-0060
Supplier:  BURKLE
undefined
All-layer scratcher
Catalog # 300-0060
Supplier:  BURKLE

Specifications

  • Ø:
    25 mm
  • Volume:
    180 ml
  • Description:
    All-layer scratcher
  • Length:
    1000 mm
  • Material:
    Stainless steel/PTFE
  • Pk:
    1

Specifications

About this item

The scratcher is designed for powders and granules that do not flow freely.

  • Sample transfer to the closable inner tube via a scraper
  • Sliding PTFE separators to divide the inner tube into different zones – also prevent the sample from being mixed

Suitable for caked bulked goods.