Order Entry
Belgium
ContactUsLinkComponent
Deep well plates, 96 square well, Bioblock™
Deep well plates, 96 square well, Bioblock™
Catalog # SIMPT110-10
Supplier:  Simport Scientific
undefined
Deep well plates, 96 square well, Bioblock™
Catalog # SIMPT110-10
Supplier:  Simport Scientific

Specifications

  • Packed:
    4 plates/bag
  • Recommended working volume:
    2,0 ml
  • Colour:
    Natural
  • Description:
    Deep 96 well plate, square wells with round bottom
  • Seq:
    01
  • W×D×H:
    127,8×85,5×41,6 mm
  • Pk:
    24
  • Well volume:
    2,2 ml

Specifications

About this item

PP. This 2,2 ml well capacity (2,1 ml when capped) plate is used mainly for compound storage and enzyme assays.

  • Conforms to SBS standards
  • Suitable to be used with Qiagen equipment
  • DMSO resistant
Delivery: Packed in a bag of 4 plates.