Orders LinkContactUsLinkComponent
Flat optical windows
Flat optical windows
Supplier:   Hellma
Flat optical windows
Supplier:  Hellma
Certificates
Flat optical windows
Flat optical windows
5 Options Available Below
Product Details & Documents
Optical glass cells and elements dedicated to special applications with detachable window.
Suitable for high precision analytical techniques.
Product Family Options
Product Information
  • Window material
    Dimensions
    Thickness
    Availability
    Price
  • HELL665-000-40
    Flat optics rectangular
    Flat optics rectangular
    Quartz (Suprasil®)
    12,5 × 45 mm
    1,25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 40,60 €
    40,60 €
    1 * 1 ST
       40,60 €
  • HELL201-1-23
    Flat optics ring for spring clap
    Flat optics ring for spring clap
    Duran glass
    21 mm
    1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 120,00 €
    120,00 €
    1 * 1 ST
       120,00 €
  • HELL202-40
    Flat optics disc
    Flat optics disc
    Quartz (Suprasil®)
    22 mm
    1,25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 42,30 €
    42,30 €
    1 * 1 ST
       42,30 €
  • HELL665-000-46
    Flat optics rectangular
    Flat optics rectangular
    Quartz (Suprasil® 300)
    12,5 × 45 mm
    1,25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 66,00 €
    66,00 €
    1 * 1 ST
       66,00 €
  • HELL202-46
    Flat optics disc
    Flat optics disc
    Quartz (Suprasil® 300)
    22 mm
    1,25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 64,90 €
    64,90 €
    1 * 1 ST
       64,90 €