Order Entry
Belgium
Orders LinkContactUsLinkComponent
3-Nitrophenol

3-Nitrophenol

Glass bottle for solids, 25 g
Catalog # MOLE50430656-25G
Toxic
Marketsource
Supplier:   Molekula
3-Nitrophenol
Catalog # MOLE50430656-25G
Supplier:  Molekula
Marketsource
CAS Number:  554-84-7
3-Nitrophenol

Glass bottle for solids, 25 g

CAS Number:  554-84-7
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  m-Nitrophenol
  • CAS Number:554-84-7
  • Formula:C₆H₅NO₃
  • MDL Number:MFCD00007240
  • Molecular Weight:139.11 g/mol
  • Merck Index:12,06682
  • Boiling Pt:265 °C (1013 hPa)
  • Melting Pt:32…35 °C
  • Density:1.37 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:209-079-8
  • UN:3458
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications