Orders LinkContactUsLinkComponent
Pipette tips
Supplier:   KARTELL
Pipette tips
Supplier:  KARTELL
Certificates
Pipette tips
47 Options Available Below
Product Details & Documents
These tips are recommended for use with the Kartell automatic micropipettes but deliver excellent results even if used with the most other micropipette brands available on the market.
  • High clarity and excellent water-repellent properties designed to keep sample retention within the tip to a minimum
  • Precise centering of the orifice at the end of the tip guarantees reliable directionality of the sample when delivered
  • Tips in racks come with snap-on lid
  • Boxes containing 'macro' tips (5000 μl and 10000 μl) have a sliding horizontal cover
  • Containers and tips are autoclavable to +121 °C for 20 minutes
Product Family Options
Product Information
  • Tip style
    Volume
    Sterile
    Tip colour
    For use with
    Packed
    Availability
    Price
  • Standard
    0,1 - 10 µl
    -
    Transparent
    Gilson® P2, P10
    Bulk
    Log in for availability
    Log in for availability
    1 * 20.000 ST
    1 * 20.000 ST - 1 460,00 €
    1 460,00 €
    1 * 20.000 ST
       1 460,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    KART0095600
  • Standard
    0,1 - 10 µl
    -
    Transparent
    Gilson® P2, P10
    Rack
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 209,00 €
    209,00 €
    1 * 960 ST
       209,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    KART9646
  • Standard
    0,1 - 10 µl
    -
    Transparent
    Gilson® P2, P10
    Refill pack
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - 77,10 €
    77,10 €
    1 * 480 ST
       77,10 €
    Specifications:
    More
    Specifications
    Cat. No.
    KART9747
  • Eppendorf® cristall
    0,5 - 10 µl
    -
    Transparent
    Kartell® Eppendorf® Socorex®, Hamilton
    Rack
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 138,00 €
    138,00 €
    1 * 960 ST
       138,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    KART9624
  • Eppendorf® cristall
    0,5 - 20 µl
    -
    Transparent
    Kartell® Pluripet PL10, Eppendorf®, Socorex®, Hamilton®
    Bulk
    Log in for availability
    Log in for availability
    1 * 25.000 ST
    1 * 25.000 ST - 767,00 €
    767,00 €
    1 * 25.000 ST
       767,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    KART0095900
  • Eppendorf® cristall
    0,5 - 20 µl
    -
    Transparent
    Kartell® Pluripet PL10, Eppendorf®, Socorex®, Hamilton®
    Rack
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 200,00 €
    200,00 €
    1 * 960 ST
       200,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    KART9650
  • Eppendorf® cristall
    0,5 - 20 µl
    -
    Transparent
    Kartell® Pluripet PL10, Eppendorf®, Socorex®, Hamilton®
    Refill pack
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - 70,90 €
    70,90 €
    1 * 480 ST
       70,90 €
    Specifications:
    More
    Specifications
    Cat. No.
    KART9746
  • Standard
    2 - 20 µl
    -
    Transparent
    Eppendorf®, Gilson®
    Rack
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 183,00 €
    183,00 €
    1 * 960 ST
       183,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    KART9657
  • Standard
    2 - 20 µl
    -
    Transparent
    Eppendorf®, Gilson®
    Bulk
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 26,00 €
    26,00 €
    1 * 1.000 ST
       26,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    KART979
  • Standard
    2 - 20 µl
    -
    Transparent
    Eppendorf®, Gilson®
    Refill pack
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - 66,80 €
    66,80 €
    1 * 480 ST
       66,80 €
    Specifications:
    More
    Specifications
    Cat. No.
    KART9745
  • Standard
    2 - 200 µl
    -
    Yellow
    Gilson®, Eppendorf®, Socorex®, Nichiryo®, Biohit®, Brand®, Hamilton®, Kartell®
    Bulk
    Log in for availability
    Log in for availability
    1 * 24.000 ST
    1 * 24.000 ST - 413,00 €
    413,00 €
    1 * 24.000 ST
       413,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    KART0096006
  • Graduated
    2 - 200 µl
    -
    Yellow
    Gilson®, Eppendorf®, Socorex®, Nichiryo®, Biohit®, Brand®, Hamilton®, Kartell®
    Bulk
    Log in for availability
    Log in for availability
    1 * 24.000 ST
    1 * 24.000 ST - 425,00 €
    425,00 €
    1 * 24.000 ST
       425,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    KART0098006
  • Standard
    2 - 200 µl
    -
    Yellow
    Gilson®
    Rack
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 109,00 €
    109,00 €
    1 * 960 ST
       109,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    KART9585
  • Standard
    2 - 200 µl
    -
    Yellow
    Kartell, Eppendorf®, Socorex, Finnpipette®, Brand, Hamilton
    Rack
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 93,20 €
    93,20 €
    1 * 960 ST
       93,20 €
    Specifications:
    More
    Specifications
    Cat. No.
    KART9605
  • Standard
    2 - 300 µl
    -
    Transparent
    Gilson®, Sartorius, Finnpipette®, Labsystem®
    Rack
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 183,00 €
    183,00 €
    1 * 960 ST
       183,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    KART9644
  • Standard
    2 - 300 µl
    -
    Transparent
    Gilson®, Sartorius, Finnpipette®, Labsystem®
    Bulk
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 48,40 €
    48,40 €
    1 * 1.000 ST
       48,40 €
    Specifications:
    More
    Specifications
    Cat. No.
    KART971
  • Standard
    2 - 300 µl
    -
    Transparent
    Gilson®, Sartorius, Finnpipette®, Labsystem®
    Refill pack
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - 67,90 €
    67,90 €
    1 * 480 ST
       67,90 €
    Specifications:
    More
    Specifications
    Cat. No.
    KART9752
  • Graduated
    5 - 200 µl
    -
    Yellow
    Gilson® P20, P50, P100, P200
    Bulk
    Log in for availability
    Log in for availability
    1 * 25.000 ST
    1 * 25.000 ST - 712,00 €
    712,00 €
    1 * 25.000 ST
       712,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    KART940
  • Standard
    5 - 200 µl
    -
    Yellow
    Gilson®, Kartell® Multipet
    Bulk
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 22,00 €
    22,00 €
    1 * 1.000 ST
       22,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    KART958
  • Standard
    5 - 200 µl
    -
    Transparent
    MLA®
    Bulk
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 31,70 €
    31,70 €
    1 * 1.000 ST
       31,70 €
    Specifications:
    More
    Specifications
    Cat. No.
    KART968
  • Standard
    5 - 200 µl
    -
    Yellow
    Kartell®, Eppendorf®, Gilson® , Biohit® , Brand® , Socorex®, HTL®, Nichiryo®, Finnpipette®, Thermo®
    Rack
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 175,00 €
    175,00 €
    1 * 960 ST
       175,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    KART9641
  • Graduated
    5 - 200 µl
    -
    Yellow
    Gilson® P20, P50, P100, P200
    Rack
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 183,00 €
    183,00 €
    1 * 960 ST
       183,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    KART9643
  • Standard
    5 - 200 µl
    -
    Yellow
    Kartell®, Eppendorf®, Gilson® , Biohit® , Brand® , Socorex®, HTL®, Nichiryo®, Finnpipette®, Thermo®
    Refill pack
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - 63,40 €
    63,40 €
    1 * 480 ST
       63,40 €
    Specifications:
    More
    Specifications
    Cat. No.
    KART9749
  • Graduated
    5 - 200 µl
    -
    Yellow
    Gilson®, Kartell® multipet
    Refill pack
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - 67,90 €
    67,90 €
    1 * 480 ST
       67,90 €
    Specifications:
    More
    Specifications
    Cat. No.
    KART9751
  • Graduated
    10 - 300 µl
    -
    Transparent
    Eppendorf®
    Bulk
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 43,00 €
    43,00 €
    1 * 1.000 ST
       43,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    KART935
  • Graduated
    10 - 300 µl
    -
    Transparent
    Eppendorf®
    Rack
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 183,00 €
    183,00 €
    1 * 960 ST
       183,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    KART9647
  • Standard
    10 - 300 µl
    -
    Transparent
    Eppendorf®
    Refill pack
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - 66,80 €
    66,80 €
    1 * 480 ST
       66,80 €
    Specifications:
    More
    Specifications
    Cat. No.
    KART9754
  • Standard
    100 - 1000 µl
    -
    Blue
    Gilson® P500, P1000
    Bulk
    Log in for availability
    Log in for availability
    1 * 10.000 ST
    1 * 10.000 ST - 391,00 €
    391,00 €
    1 * 10.000 ST
       391,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    KART941
  • Standard
    100 - 1000 µl
    -
    Blue
    Kartell, Eppendorf, Gilson, Socorex, Brand, Nichiryo®, Sartorius, Hamilton
    Bulk
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 22,20 €
    22,20 €
    1 * 1.000 ST
       22,20 €
    Specifications:
    More
    Specifications
    Cat. No.
    612-1418
  • Standard
    100 - 1000 µl
    -
    Blue
    Kartell, Eppendorf, Gilson, Socorex, Finnpipette, Brand
    Rack
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 122,00 €
    122,00 €
    1 * 960 ST
       122,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    KART9615
  • Standard
    100 - 1000 µl
    -
    Blue
    Kartell, Eppendorf, Gilson, Socorex, Brand, Nichiryo®, Sartorius, Hamilton
    Rack
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 179,00 €
    179,00 €
    1 * 960 ST
       179,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    KART9640
  • Standard
    100 - 1000 µl
    -
    Blue
    Gilson® P500, P1000
    Rack
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 200,00 €
    200,00 €
    1 * 960 ST
       200,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    KART9645
  • Standard
    100 - 1000 µl
    -
    Blue
    Kartell, Eppendorf, Gilson, Socorex, Brand, Nichiryo®, Sartorius, Hamilton
    Refill pack
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - 66,40 €
    66,40 €
    1 * 480 ST
       66,40 €
    Specifications:
    More
    Specifications
    Cat. No.
    KART9748
  • Standard
    100 - 1000 µl
    -
    Blue
    Gilson® P500, P1000
    Refill pack
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - 68,70 €
    68,70 €
    1 * 480 ST
       68,70 €
    Specifications:
    More
    Specifications
    Cat. No.
    KART9753
  • Standard
    200 - 1000 µl
    -
    Blue
    Kartell®, Gilson®, Eppendorf®, Sartorius
    Bulk
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 39,30 €
    39,30 €
    1 * 1.000 ST
       39,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    KART957
  • Standard
    200 - 1000 µl
    -
    Transparent
    MLA®
    Bulk
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 44,40 €
    44,40 €
    1 * 1.000 ST
       44,40 €
    Specifications:
    More
    Specifications
    Cat. No.
    KART969
  • Standard, green
    250 - 1000 µl
    -
    Transparent
    Oxford® (old model)
    Bulk
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 44,40 €
    44,40 €
    1 * 1.000 ST
       44,40 €
    Specifications:
    More
    Specifications
    Cat. No.
    KART963
  • Standard
    250 - 1000 µl
    -
    Transparent
    Beckman®, Sherwood®, Lancer®
    Bulk
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 47,40 €
    47,40 €
    1 * 1.000 ST
       47,40 €
    Specifications:
    More
    Specifications
    Cat. No.
    613-0500
  • Standard
    250 - 1000 µl
    -
    Blue
    Oxford® (slim line)
    Bulk
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 44,00 €
    44,00 €
    1 * 1.000 ST
       44,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    KART967
  • Standard
    1000 - 5000 µl
    -
    Transparent
    Finnpipette®, Treff, Brand
    Bulk
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 527,00 €
    527,00 €
    1 * 2.000 ST
       527,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    KART942
  • Standard
    1000 - 5000 µl
    -
    Transparent
    Kartell®, Eppendorf®, Biohit®, Socorex® , HTL®
    Bulk
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 162,00 €
    162,00 €
    1 * 250 ST
       162,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    KART964
  • Standard
    1000 - 5000 µl
    -
    Blue
    Kartell® Pluripet std and Pluripet II, Gilson®, Socorex®, Nichiryo®
    Bulk
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 70,10 €
    70,10 €
    1 * 250 ST
       70,10 €
    Specifications:
    More
    Specifications
    Cat. No.
    KART984
  • Standard
    1000 - 5000 µl
    -
    Blue
    Kartell® Pluripet std and Pluripet II, Gilson®, Socorex®, Nichiryo®
    Rack
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 208,00 €
    208,00 €
    1 * 200 ST
       208,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    KART9648
  • Standard
    1000 - 5000 µl
    -
    Transparent
    Finnpipette®, Treff, Brand
    Rack
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 13,70 €
    13,70 €
    1 * 50 ST
       13,70 €
    Specifications:
    More
    Specifications
    Cat. No.
    KART9847
  • Standard
    1000 - 5000 µl
    -
    Transparent
    Kartell®, Eppendorf®, Biohit®, Socorex® , HTL®
    Rack
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 208,00 €
    208,00 €
    1 * 4 ST
       208,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    KART0964900
  • Standard
    2000 - 10000 µl
    -
    Transparent
    Gilson®, Socorex®, Labsystem®, Finnpipette®
    Bulk
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 626,00 €
    626,00 €
    1 * 1.000 ST
       626,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    KART943
  • Standard
    2000 - 10000 µl
    -
    Transparent
    Gilson®, Socorex®, Labsystem®, Finnpipette®
    Rack
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 247,00 €
    247,00 €
    1 * 100 ST
       247,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    KART9848