Orders LinkContactUsLinkComponent
Burettes, straight stopcock, PYREX®
Burettes, straight stopcock, PYREX®
Supplier:   SciLabware
Burettes, straight stopcock, PYREX®
Supplier:  SciLabware
Certificates
Burettes, straight stopcock, PYREX®
Burettes, straight stopcock, PYREX®
8 Options Available Below
Product Details & Documents
Borosilicate glass with interchangeable PTFE key, Class AS.
  • Calibrated to deliver ‘Ex’
  • PTFE key for grease-free perfomance
  • White enamel graduations for good visibility with dark coloured solutions
Certifications: ISO 385.
Documents
Product Family Options
Class AS burettes
Product Information
  • Capacity
    Division
    Tolerance
    Availability
    Price
  • 10 ml
    0,02 ml
    0,02 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 48,60 €
    48,60 €
    1 * 1 ST
       48,60 €
  • 25 ml
    0,10 ml
    0,05 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 40,90 €
    40,90 €
    1 * 1 ST
       40,90 €
  • 50 ml
    0,10 ml
    0,05 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 42,40 €
    42,40 €
    1 * 1 ST
       42,40 €
  • 100 ml
    0,20 ml
    0,10 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 56,30 €
    56,30 €
    1 * 1 ST
       56,30 €
Class AS burettes, works certified
Product Information
  • Capacity
    Division
    Tolerance
    Availability
    Price
  • 10 ml
    0,02 ml
    0,02 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 81,60 €
    81,60 €
    1 * 1 ST
       81,60 €
  • 25 ml
    0,10 ml
    0,05 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 58,90 €
    58,90 €
    1 * 1 ST
       58,90 €
  • 50 ml
    0,10 ml
    0,05 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 58,90 €
    58,90 €
    1 * 1 ST
       58,90 €
  • 100 ml
    0,20 ml
    0,10 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 99,40 €
    99,40 €
    1 * 1 ST
       99,40 €