ClipTip™ Pipette Tips
About this item
ClipTip™ pipette tips provide security with a unique and innovative interlocking technology that ensures a complete seal on every channel with minimal tip attachment and ejection force.
- Pipette tips feature 'clips' that lock tips in place and forms a complete seal
- Every ClipTip™ pipette tip utilises low retention technology to enable maximium sample recovery
- Certified free of RNase, DNase, DNA, ATP and endotoxin contamination
- Each tip insert has ink-jetted lot and volume information; the convenient colour key makes it easy to match the colour coding of the F1-ClipTip™ and E1-ClipTip™ pipettes with the corresponding ClipTip™ pipette tip rack insert
- Environmentally friendly reload system uses recycled components and 30% less plastic
Achieve newfound confidence knowing that once attached, your tips are locked firmly in place, and will not loosen or fall off regardless of application pressure.
ClipTip™ pipette tips feature 'clips' that lock tips in place and form a complete seal, Each time ClipTip pipette tips are on, they are sealed and will not loosen regardless of application pressure, ClipTip™ pipette tips are clipped securely on the F1-ClipTip™ and E1-ClipTip™ pipettes with a light touch eliminating the frustration of banging tips on your pipette, Low retention technology minimizes liquid retention, and enable maximium sample recovery.
40 Options Available Below
Product Details & Documents
ClipTip™ pipette tips provide security with a unique and innovative interlocking technology that ensures a complete seal on every channel with minimal tip attachment and ejection force.
- Pipette tips feature 'clips' that lock tips in place and forms a complete seal
- Every ClipTip™ pipette tip utilises low retention technology to enable maximium sample recovery
- Certified free of RNase, DNase, DNA, ATP and endotoxin contamination
- Each tip insert has ink-jetted lot and volume information; the convenient colour key makes it easy to match the colour coding of the F1-ClipTip™ and E1-ClipTip™ pipettes with the corresponding ClipTip™ pipette tip rack insert
- Environmentally friendly reload system uses recycled components and 30% less plastic
Achieve newfound confidence knowing that once attached, your tips are locked firmly in place, and will not loosen or fall off regardless of application pressure.
ClipTip™ pipette tips feature 'clips' that lock tips in place and form a complete seal, Each time ClipTip pipette tips are on, they are sealed and will not loosen regardless of application pressure, ClipTip™ pipette tips are clipped securely on the F1-ClipTip™ and E1-ClipTip™ pipettes with a light touch eliminating the frustration of banging tips on your pipette, Low retention technology minimizes liquid retention, and enable maximium sample recovery.
Documents
Certificates
Search by Lot NumberAppNote_KFDuoPrime_E1CT Eq_012016 AT-Smart Note E1-ClipTip Adjustable Tip Spacing_RevA AT-Thermo_Scientific_ClipTip_Technology_Transform Daily Pipetting-AppNote1 E1-ClipTip Tip Attachment and Ejection Forces Smart Note Thermo Scientific E1-ClipTip Gel Loading for PCR
Application Notes
BRHPCLIPEUVWR0916_V2
Product documentatie
Specifications for Product Family
Specifications
- Catalog No:
- 613-6031
- 613-6032
- 613-6025
- 613-6026
- 613-6861
- 613-6862
- 613-6192
- 613-6193
- 613-2205
- 613-2206
- 613-2207
- 613-2208
- 613-6027
- 613-6028
- 613-2209
- 613-2210
- 613-2211
- 613-2212
- 613-6029
- 613-6030
- 613-2213
- 613-2214
- 613-2215
- 613-2216
- 613-2217
- 613-2218
- 613-2219
- 613-2220
- 613-6194
- 613-6195
- 613-7447
- 613-7446
- 613-2222
- 613-2223
- 613-2224
- 613-2225
- 613-6033
- 613-6034
- 613-6035
- 613-6036
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
Specifications
Product Family Options
ClipTip 12,5
Product Information
- Tip styleVolumeSterileLengthColour codeFor use withPackedAvailabilityPrice
- Standard0,1 - 12,5 µl-30 mmPinkE1-ClipTip and F1-ClipTip pipettes10 racks, 96 eachLog in for availabilityLog in for availability1 * 960 ST1 * 960 ST - 47,00 €47,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 ST47,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-6031Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - Standard0,1 - 12,5 µl+30 mmPinkE1-ClipTip and F1-ClipTip pipettes10 racks, 96 eachLog in for availabilityLog in for availability1 * 960 ST1 * 960 ST - 61,00 €61,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 ST61,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-6032Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - Standard0,1 - 12,5 µl-30 mmPinkE1-ClipTip and F1-ClipTip pipettes10 racks, 384 eachLog in for availabilityLog in for availability1 * 3.840 ST1 * 3.840 ST - 307,00 €307,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 3.840 ST307,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-6025Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - Standard0,1 - 12,5 µl+30 mmPinkE1-ClipTip and F1-ClipTip pipettes10 racks, 384 eachLog in for availabilityLog in for availability1 * 3.840 ST1 * 3.840 ST - 361,00 €361,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 3.840 ST361,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-6026Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - Extended0,1 - 12,5 µl-41 mmPinkE1-ClipTip and F1-ClipTip pipettes10 racks, 96 eachLog in for availabilityLog in for availability1 * 960 ST1 * 960 ST - 46,00 €46,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 ST46,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-6861Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - Extended0,1 - 12,5 µl+41 mmPinkE1-ClipTip and F1-ClipTip pipettes10 racks, 96 eachLog in for availabilityLog in for availability1 * 960 ST1 * 960 ST - 60,00 €60,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 ST60,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-6862Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationIRECS; ISO 14001 (Certificate Registration No. EMS 752603)Low Manufacturing Impact - Extended0,1 - 12,5 µl-41 mmPinkE1-ClipTip and F1-ClipTip pipettes10 racks, 384 eachLog in for availabilityLog in for availability1 * 3.840 ST1 * 3.840 ST - 300,00 €300,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 3.840 ST300,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-6192Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - Extended0,1 - 12,5 µl+41 mmPinkE1-ClipTip and F1-ClipTip pipettes10 racks, 384 eachLog in for availabilityLog in for availability1 * 3.840 ST1 * 3.840 ST - 354,00 €354,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 3.840 ST354,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-6193Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact
ClipTip 20
Product Information
- Tip styleVolumeSterileLengthColour codeFor use withPackedAvailabilityPrice
- Low Retention1 - 20 µl-46 mmPinkF1-ClipTip pipettes10 racks, 96 eachLog in for availabilityLog in for availability1 * 960 ST1 * 960 ST - 47,00 €47,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 ST47,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-2205Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - Low Retention1 - 20 µl+46 mmPinkF1-ClipTip pipettes10 racks, 96 eachLog in for availabilityLog in for availability1 * 960 ST1 * 960 ST - 61,00 €61,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 ST61,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-2206Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - Low Retention1 - 20 µl-46 mmPinkF1-ClipTip pipettes10 reload inserts, 96 eachLog in for availabilityLog in for availability1 * 960 ST1 * 960 ST - 44,00 €44,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 ST44,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-2207Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - Low Retention1 - 20 µl+46 mmPinkF1-ClipTip pipettes10 reload inserts, 96 eachLog in for availabilityLog in for availability1 * 960 ST1 * 960 ST - 56,00 €56,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 ST56,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-2208Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact
ClipTip 30
Product Information
- Tip styleVolumeSterileLengthColour codeFor use withPackedAvailabilityPrice
- Standard 3841 - 30 µl-44 mmVioletE1-ClipTip pipettes10 racks, 384 eachLog in for availabilityLog in for availability1 * 3.840 ST1 * 3.840 ST - 307,00 €307,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 3.840 ST307,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-6027Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - Standard 3841 - 30 µl+44 mmVioletE1-ClipTip pipettes10 racks, 384 eachLog in for availabilityLog in for availability1 * 3.840 ST1 * 3.840 ST - 361,00 €361,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 3.840 ST361,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-6028Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact
ClipTip 50
Product Information
- Tip styleVolumeSterileLengthColour codeFor use withPackedAvailabilityPrice
- Low Retention5 - 50 µl-51 mmVioletF1-ClipTip pipettes10 racks, 96 eachLog in for availabilityLog in for availability1 * 960 ST1 * 960 ST - 47,00 €47,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 ST47,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-2209Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - Low Retention5 - 50 µl+51 mmVioletF1-ClipTip pipettes10 racks, 96 eachLog in for availabilityLog in for availability1 * 960 ST1 * 960 ST - 60,00 €60,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 ST60,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-2210Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - Low Retention5 - 50 µl-51 mmVioletF1-ClipTip pipettes10 reload inserts, 96 eachLog in for availabilityLog in for availability1 * 960 ST1 * 960 ST - 44,00 €44,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 ST44,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-2211Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - Low Retention5 - 50 µl+51 mmVioletF1-ClipTip pipettes10 reload inserts, 96 eachLog in for availabilityLog in for availability1 * 960 ST1 * 960 ST - 56,00 €56,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 ST56,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-2212Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact
ClipTip 125
Product Information
- Tip styleVolumeSterileLengthColour codeFor use withPackedAvailabilityPrice
- Standard 3841 - 125 µl-64 mmYellowE1-ClipTip pipettes10 racks, 384 eachLog in for availabilityLog in for availability1 * 3.840 ST1 * 3.840 ST - 301,00 €301,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 3.840 ST301,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-6029Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - Standard 3841 - 125 µl+64 mmYellowE1-ClipTip pipettes10 racks, 384 eachLog in for availabilityLog in for availability1 * 3.840 ST1 * 3.840 ST - 355,00 €355,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 3.840 ST355,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-6030Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact
ClipTip 200
Product Information
- Tip styleVolumeSterileLengthColour codeFor use withPackedAvailabilityPrice
- Low Retention10 - 200 µl-56 mmYellowE1-ClipTip and F1-ClipTip pipettes10 racks, 96 eachLog in for availabilityLog in for availability1 * 960 ST1 * 960 ST - 47,00 €47,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 ST47,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-2213Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - Low Retention10 - 200 µl+56 mmYellowE1-ClipTip and F1-ClipTip pipettes10 racks, 96 eachLog in for availabilityLog in for availability1 * 960 ST1 * 960 ST - 60,00 €60,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 ST60,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-2214Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - Low Retention10 - 200 µl-56 mmYellowE1-ClipTip and F1-ClipTip pipettes10 reload inserts, 96 eachLog in for availabilityLog in for availability1 * 960 ST1 * 960 ST - 44,00 €44,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 ST44,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-2215Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - Low Retention10 - 200 µl+56 mmYellowE1-ClipTip and F1-ClipTip pipettes10 reload inserts, 96 eachLog in for availabilityLog in for availability1 * 960 ST1 * 960 ST - 56,00 €56,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 ST56,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-2216Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact
ClipTip 300
Product Information
- Tip styleVolumeSterileLengthColour codeFor use withPackedAvailabilityPrice
- Low Retention30 - 300 µl-63 mmOrangeE1-ClipTip and F1-ClipTip pipettes10 racks, 96 eachLog in for availabilityLog in for availability1 * 960 ST1 * 960 ST - 47,00 €47,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 ST47,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-2217Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - Low Retention30 - 300 µl+63 mmOrangeE1-ClipTip and F1-ClipTip pipettes10 racks, 96 eachLog in for availabilityLog in for availability1 * 960 ST1 * 960 ST - 60,00 €60,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 ST60,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-2218Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - Low Retention30 - 300 µl-63 mmOrangeE1-ClipTip and F1-ClipTip pipettes10 reload inserts, 96 eachLog in for availabilityLog in for availability1 * 960 ST1 * 960 ST - 44,00 €44,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 ST44,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-2219Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - Low Retention30 - 300 µl+63 mmOrangeE1-ClipTip and F1-ClipTip pipettes10 reload inserts, 96 eachLog in for availabilityLog in for availability1 * 960 ST1 * 960 ST - 56,00 €56,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 ST56,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-2220Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - Low Retention, Extended Length30 - 300 µl-100 mmOrangeE1-ClipTip and F1-ClipTip pipettes8 racks, 96 eachLog in for availability1 * 768 ST47,90 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.613-6194Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - Low Retention, Extended Length30 - 300 µl+100 mmOrangeE1-ClipTip and F1-ClipTip pipettes8 racks, 96 eachLog in for availabilityLog in for availability1 * 768 ST1 * 768 ST - 50,00 €50,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 768 ST50,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-6195Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - Low Retention, Extended Length30 - 300 µl+100 mmOrangeE1-ClipTip and F1-ClipTip pipettes10 reload inserts, 96 eachLog in for availabilityLog in for availability1 * 960 ST1 * 960 ST - 58,00 €58,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 ST58,00 €Specifications:More
Specifications
Cat. No.613-7447Environmentally PreferableEnvironmental certification - Low Retention, Extended Length30 - 300 µl-100 mmOrangeE1-ClipTip and F1-ClipTip pipettes10 reload inserts, 96 eachLog in for availabilityLog in for availability1 * 960 ST1 * 960 ST - 45,00 €45,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 ST45,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-7446Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact
ClipTip 1000
Product Information
- Tip styleVolumeSterileLengthColour codeFor use withPackedAvailabilityPrice
- Low Retention100 - 1000 µl-95 mmBlueE1-ClipTip and F1-ClipTip pipettes8 racks, 96 eachLog in for availabilityLog in for availability1 * 768 ST1 * 768 ST - 52,00 €52,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 768 ST52,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-2222Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - Low Retention100 - 1000 µl+95 mmBlueE1-ClipTip and F1-ClipTip pipettes8 racks, 96 eachLog in for availabilityLog in for availability1 * 768 ST1 * 768 ST - 66,00 €66,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 768 ST66,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-2223Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - Low Retention100 - 1000 µl-95 mmBlueE1-ClipTip and F1-ClipTip pipettes8 reload inserts, 96 eachLog in for availabilityLog in for availability1 * 768 ST1 * 768 ST - 48,00 €48,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 768 ST48,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-2224Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - Low Retention100 - 1000 µl+95 mmBlueE1-ClipTip and F1-ClipTip pipettes8 reload inserts, 96 eachLog in for availabilityLog in for availability1 * 768 ST1 * 768 ST - 61,00 €61,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 768 ST61,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-2225Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact
ClipTip 1250
Product Information
- Tip styleVolumeSterileLengthColour codeFor use withPackedAvailabilityPrice
- Low Retention15 - 1250 µl-103 mmTurquoiseE1-ClipTip pipettes8 racks, 96 eachLog in for availabilityLog in for availability1 * 768 ST1 * 768 ST - 51,00 €51,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 768 ST51,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-6033Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - Low Retention15 - 1250 µl+103 mmTurquoiseE1-ClipTip pipettes8 racks, 96 eachLog in for availabilityLog in for availability1 * 768 ST1 * 768 ST - 65,00 €65,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 768 ST65,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-6034Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - Low Retention15 - 1250 µl-103 mmTurquoiseE1-ClipTip pipettes8 reload inserts, 96 eachLog in for availability1 * 768 ST58,90 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.613-6035Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - Low Retention15 - 1250 µl+103 mmTurquoiseE1-ClipTip pipettes8 reload inserts, 96 eachLog in for availabilityLog in for availability1 * 768 ST1 * 768 ST - 59,00 €59,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 768 ST59,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-6036Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact
Racks without tips
Product Information
- Catalog #DescriptionAvailabilityPrice
- Empty rack, medium, colour code yellow and orange (for ClipTip 200 and 300)Log in for availability1 * 10 ST23,00 €