Orders LinkContactUsLinkComponent
Round slide mailer, LockMailer™
Round slide mailer, LockMailer™
Slide containers, Magenta, HDPE, 4
Catalog # SIMPM950-4MA
Supplier:   Simport Scientific
Round slide mailer, LockMailer™
Catalog # SIMPM950-4MA
Supplier:  Simport Scientific
Round slide mailer, LockMailer™
Round slide mailer, LockMailer™
Slide containers, Magenta, HDPE, 4
Price
513,00 €
Log in for availability
1 * 500 ST
Product Details & Documents
Transparent container made of high density polyethylene for mailing or storing slides between the doctor's office and the lab.
  • Inside channels slotted to keep slides separated
  • Unique tamper evident leakproof screw cap
  • Good stability on a bench
Specifications
  • No. of slides:
    4
  • Colour:
    Magenta
  • Description:
    Slide containers
  • Material:
    HDPE