Draining racks, with drip pan
Supplier: BOCHEM
Jump to:
About this item
These PA coated wire draining racks are suitable for test tubes and flasks.
- With drip pan
- Good chemical resistance
- Good temperature resistance
2 Options Available Below
Product Details & Documents
These PA coated wire draining racks are suitable for test tubes and flasks.
- With drip pan
- Good chemical resistance
- Good temperature resistance
Product Family Options
Product Information
- W×D×HAvailabilityPrice
- BOCH10140Draining rack, with 24 rods (95 mm) and 20 arches (2×110 mm and 3×70 mm)Draining rack, with 24 rods (95 mm) and 20 arches (2×110 mm and 3×70 mm)420×160×610 mmLog in for availability1 * 1 ST199,00 €
- BOCH10130Draining rack, wth 60 rods (95 mm) and 5 arches (2×110 mm and 3×70 mm)Draining rack, wth 60 rods (95 mm) and 5 arches (2×110 mm and 3×70 mm)420×160×610 mmLog in for availability1 * 1 ST167,00 €