Orders LinkContactUsLinkComponent
Watch dishes
Watch dishes
Supplier:   KARTELL
Watch dishes
Supplier:  KARTELL
Certificates
Watch dishes
5 Options Available Below
Product Details & Documents
Made of PTFE, white.
  • Very good chemical, heat and mechanical resistance
  • Heat resistant up to +120 °C
  • For use with foodstuff
For blends or for covering vessels.
Product Family Options
Product Information
  • Ø
    Availability
    Price
  • KART643
    Watch glass, PTFE
    Watch glass, PTFE
    52,76 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 17,10 €
    17,10 €
    1 * 1 ST
       17,10 €
  • KART644
    Watch glass, PTFE
    Watch glass, PTFE
    76,32 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 26,00 €
    26,00 €
    1 * 1 ST
       26,00 €
  • KART645
    Watch glass, PTFE
    Watch glass, PTFE
    99,39 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 39,00 €
    39,00 €
    1 * 1 ST
       39,00 €
  • KART646
    Watch glass, PTFE
    Watch glass, PTFE
    126,36 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 52,00 €
    52,00 €
    1 * 1 ST
       52,00 €
  • KART647
    Watch glass, PTFE
    Watch glass, PTFE
    153,92 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 66,30 €
    66,30 €
    1 * 1 ST
       66,30 €