Test tubes with screw cap
Supplier: KARTELL
Jump to:
About this item
PS, clear, with PE screw cap.
- Good thermal resistance
- Good chemical resistance
Tubes have a round bottom and a freestanding skirt. Sterile tubes are available individually wrapped. The 30 ml individual tubes have a self-adhesive label for marking.
4 Options Available Below
Product Details & Documents
PS, clear, with PE screw cap.
- Good thermal resistance
- Good chemical resistance
Tubes have a round bottom and a freestanding skirt. Sterile tubes are available individually wrapped. The 30 ml individual tubes have a self-adhesive label for marking.
Product Family Options
Product Information
- CapacityLengthExt.ØSterilityAvailabilityPrice
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.