Orders LinkContactUsLinkComponent
Dispenser bottles, Wheaton®
Dispenser bottles, Wheaton®
Supplier:   DWK Life Sciences
Dispenser bottles, Wheaton®
Supplier:  DWK Life Sciences
Certificates
Dispenser bottles, Wheaton®
Dispenser bottles, Wheaton®
2 Options Available Below
Product Details & Documents
These translucent dispensing bottles are excellent for mild and strong buffers with a spout to dispense the contents and then could be depressed to seal the bottle.
  • Very flexible with high impact strength
  • Good chemical resistance
Product Family Options
Product Information
  • Capacity
    Material
    Height
    Ext.Ø
    Thread
    Availability
    Price
  • 60 ml
    LDPE
    92 mm
    39 mm
    20-410
    Log in for availability
    Log in for availability
    1 * 144 ST
    1 * 144 ST - 455,00 €
    455,00 €
    1 * 144 ST
       455,00 €
  • 125 ml
    LDPE
    130 mm
    43 mm
    20-410
    Log in for availability
    Log in for availability
    1 * 72 ST
    1 * 72 ST - 254,00 €
    254,00 €
    1 * 72 ST
       254,00 €