Orders LinkContactUsLinkComponent
Scoops, PharmaScoop
Scoops, PharmaScoop
Supplier:   BURKLE
Scoops, PharmaScoop
Supplier:  BURKLE
Certificates
Scoops, PharmaScoop
Scoops, PharmaScoop
9 Options Available Below
Product Details & Documents
Stainless steel AISI 316 (1,4401). This sample scoop has been developed in order to meet the high requirements of the pharmaceutical industry, such as those contained in the FDA regulations.
  • GMP conform
  • Highly polished
  • Without edges, crevices or undercuts in which sample residues could collect and contaminate subsequent samples
The scoops meet all the requirements for GMP compliance in production and sampling.
Product Family Options
Product Information
  • Capacity
    Length
    Ø
    Availability
    Price
  • 10 ml
    45 mm
    25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 108,00 €
    108,00 €
    1 * 1 ST
       108,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    BURK5324-0010
  • 50 ml
    70 mm
    38 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 182,00 €
    182,00 €
    1 * 1 ST
       182,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    231-0496
  • 100 ml
    100 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 247,00 €
    247,00 €
    1 * 1 ST
       247,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    300-0148
  • 500 ml
    140 mm
    95 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 248,00 €
    248,00 €
    1 * 1 ST
       248,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    300-0154
  • 750 ml
    210 mm
    95 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 256,00 €
    256,00 €
    1 * 1 ST
       256,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    300-0149
  • 1000 ml
    235 mm
    110 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 283,00 €
    283,00 €
    1 * 1 ST
       283,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    300-0150
  • 1250 ml
    195 mm
    125 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 292,00 €
    292,00 €
    1 * 1 ST
       292,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    300-0151
  • 1700 ml
    230 mm
    135 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 303,00 €
    303,00 €
    1 * 1 ST
       303,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    300-0152
  • 2500 ml
    215 mm
    185 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 362,00 €
    362,00 €
    1 * 1 ST
       362,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    300-0153
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.