Orders LinkContactUsLinkComponent
Sampling siphons
Sampling siphons
Supplier:   BURKLE
Sampling siphons
Supplier:  BURKLE
Certificates
Sampling siphons
Sampling siphons
3 Options Available Below
Product Details & Documents
Disposable plunging samplers acting like siphons.
  • Transparent plastic for better visibility
  • PE surface resists adhesion of the material
  • Minimised sample retention on the dip sampler
If strongly adhesive liquids are sampled, it is easier to dispose of the sampler than to carry out a complicated cleaning process.
Product Family Options
Product Information
  • Material
    Capacity
    Length
    Availability
    Price
  • BURK5387-0050
    Sampling siphon
    Sampling siphon
    PE
    75 ml
    500 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 16,40 €
    16,40 €
    1 * 1 ST
       16,40 €
  • BURK5387-0075
    Sampling siphon
    Sampling siphon
    PE
    175 ml
    750 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 30,90 €
    30,90 €
    1 * 1 ST
       30,90 €
  • BURK5387-0100
    Sampling siphon
    Sampling siphon
    PE
    275 ml
    1000 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 45,40 €
    45,40 €
    1 * 1 ST
       45,40 €