Sample dippers
Supplier: BURKLE
Jump to:
About this item
Plunging siphons for the simple removal of liquid samples.
- Made from transparent PP
- All-layer and point sampling
- Upper end with reflux valve, lower end with ball valve
4 Options Available Below
Product Details & Documents
Plunging siphons for the simple removal of liquid samples.
- Made from transparent PP
- All-layer and point sampling
- Upper end with reflux valve, lower end with ball valve
Documents
Product Family Options
Product Information
- ColourMaterialLengthØAvailabilityPrice
- BURK5315-0050Dip sampler for liquid samples
- BURK5315-0100Dip sampler for liquid samples
- 331-2113Dip sampler for liquid samples
- BURK5315-0300Dip sampler for liquid samples
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.