Order Entry
Belgium
Orders LinkContactUsLinkComponent
Mechanical timers

Mechanical timers

Supplier:   KARTELL
Mechanical timers
Supplier:  KARTELL
Certificates
Mechanical timers
Mechanical timers

2 Options Available Below

Product Details & Documents

Two different manual timers made of ABS.

  • Mechanical movement (accuracy 3%)
  • Timing up to 60 minutes
  • Portable and pocket size
  • Round one hand model can be rewound using one hand only
  • An alarm sounds at the end of the set time
Product Family Options

Product Information

  • Colour
    Accuracy
    Ø
    W×D×H
    Availability
    Price
  • KART900
    Clip model
    Clip model
    White
    3 %
    56,6×56,5×28 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 52,20 €
    52,20 €
    1 * 1 ST
       52,20 €

  • KART901
    One hand model
    One hand model
    White/black
    3 %
    72 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 52,20 €
    52,20 €
    1 * 1 ST
       52,20 €