WHEATON® Rubber Stoppers
About this item
Stoppers of various styles, sizes and rubber formulations.
- Manufactured from the highest quality raw materials
- Components manufactured to yield exceptional dimensional stability
- Autoclavable
Straight plug stoppers provide maximum sealing for vials with straight-wall glass finish.
Snap-on style provides maximum sealing for vials with a blow-back glass finish.
Sleeve style stoppers feature a fold-over skirt for sealing in serum finish vials without a crimp closure.
18 Options Available Below
Product Details & Documents
Stoppers of various styles, sizes and rubber formulations.
- Manufactured from the highest quality raw materials
- Components manufactured to yield exceptional dimensional stability
- Autoclavable
Straight plug stoppers provide maximum sealing for vials with straight-wall glass finish.
Snap-on style provides maximum sealing for vials with a blow-back glass finish.
Sleeve style stoppers feature a fold-over skirt for sealing in serum finish vials without a crimp closure.
Specifications for Product Family
Specifications
- Catalog No:
- 215-4708
- 215-4709
- 215-4713
- 548-2050
- 215-4715
- 215-3481
- 215-3482
- 215-3484
- 215-3485
- 215-3486
- 215-3487
- 215-3490
- 215-3492
- 215-4712
- 215-4714
- 215-4716
- 215-4717
- 215-4718
- Durometer hardness:
- 39
- 39
- 39
- 39
- 39
- 40
- 40
- 45
- 40
- 40
- 55
- 50
- 46
- 40
- 45s
- 55
- 46
- 52
Specifications
Product Family Options
Sleeve style
Product Information
- Ø bottomØ topAvailabilityPrice
- 215-4708Stopper, natural red rubberSpecifications:
More
Specifications
Cat. No.215-4708Durometer hardness39 - 215-4709Stopper, natural red rubberSpecifications:
More
Specifications
Cat. No.215-4709Durometer hardness39 - 215-4713Stopper, natural red rubberSpecifications:
More
Specifications
Cat. No.215-4713Durometer hardness39 - 548-2050Stopper, natural red rubberSpecifications:
More
Specifications
Cat. No.548-2050Durometer hardness39 - 215-4715Stopper, natural red rubberSpecifications:
More
Specifications
Cat. No.215-4715Durometer hardness39
Straight plug style
Product Information
- Ø bottomØ topAvailabilityPrice
- 215-3481Stopper, grey chlorobutyl-isoprene blend/FEP facedStopper, grey chlorobutyl-isoprene blend/FEP faced7,0 mm13,0 mmLog in for availability1 * 1.000 ST1 330,00 €Specifications:
More
Specifications
Cat. No.215-3481Durometer hardness40 - 215-3482Stopper, natural red rubberSpecifications:
More
Specifications
Cat. No.215-3482Durometer hardness40 - 215-3484Stopper, natural red rubberSpecifications:
More
Specifications
Cat. No.215-3484Durometer hardness45 - 215-3485Stopper, grey chlorobutyl-isoprene blendSpecifications:
More
Specifications
Cat. No.215-3485Durometer hardness40 - 215-3486Stopper, grey chlorobutyl-isoprene blend/FEP facedStopper, grey chlorobutyl-isoprene blend/FEP faced13,0 mm20,0 mmLog in for availability1 * 1.000 ST1 730,00 €Specifications:
More
Specifications
Cat. No.215-3486Durometer hardness40 - 215-3487Stopper, siliconeSpecifications:
More
Specifications
Cat. No.215-3487Durometer hardness55 - 215-3490Stopper, grey bromobutylSpecifications:
More
Specifications
Cat. No.215-3490Durometer hardness50 - 215-3492Stopper, grey chlorobutylSpecifications:
More
Specifications
Cat. No.215-3492Durometer hardness46
Snap-On style
Product Information
- Ø bottomØ topAvailabilityPrice
- 215-4712Stopper, natural red rubberSpecifications:
More
Specifications
Cat. No.215-4712Durometer hardness40 - 215-4714Stopper, natural red rubberSpecifications:
More
Specifications
Cat. No.215-4714Durometer hardness45s - 215-4716Stopper, grey chlorobutylSpecifications:
More
Specifications
Cat. No.215-4716Durometer hardness55 - 215-4717Stopper, grey chlorobutylSpecifications:
More
Specifications
Cat. No.215-4717Durometer hardness46 - 215-4718Stopper, grey chlorobutylSpecifications:
More
Specifications
Cat. No.215-4718Durometer hardness52
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



