Tubing, silicone, K70
: KARTELL
Silicone, translucent.
- Excellent temperature resistance
- Autoclavable (121 °C)
- Limited chemical resistance when used with concentrated acids
For laboratory application.
Temperature resistance: –60 to +200 °C
: FDA, USP, Class VI, A 50p60, EPC VI.1.3.2 approved. Flexible and transparent.
Frequently Bought Together