Order Entry
Belgium
Orders LinkContactUsLinkComponent
Spatulas, micro

Spatulas, micro

150 mm, 7 mm
Catalog # RSGA316.150
Supplier:   RSG
Spatulas, micro
Catalog # RSGA316.150
Supplier:  RSG
Spatulas, micro
Spatulas, micro

150 mm, 7 mm

Price
59,40 €
Log in for availability
1 * 10 ST
Product Details & Documents

Stainless steel 18/10. Micro spatula with one square end.

Specifications
  • Length:
    150 mm
  • Blade width:
    7 mm