
Methyl orange ACS, Reag. Ph. Eur.
Supplier: Merck
Certificates
About this item
CAS Number:547-58-0
:
Formula:C₁₄H₁₄N₃NaO₃S
MDL Number:MFCD00007502
:327.34 g/mol
Merck Index:13,06127
Boiling Pt:∼90 °C (1013 hPa)
Melting Pt:> 300 °C
Density:∼1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:208-925-3
UN:3143
ADR:6.1,III
:M-Clarity™ quality level = MQ200
Flash Pt:38 °C
:GHS06
Specification Test Results
- Solubility:5 g/l (20 °C)
- Melting Point:300 °C
- Molar Mass:327.34 g/mol
- Bulk Density:200 - 400 kg/m3
- pH:6.5 (5 g/l, H2O, 20 °C)
- Hazard Class:6.1
- Hazard Class Description:Toxic Materials
Specification Test Results
Related Information
Frequently Bought Together