Order Entry
Belgium
Orders LinkContactUsLinkComponent
4-Fluorophenol ≥99%

4-Fluorophenol ≥99%

Corrosif
Irritant
4-Fluorophenol ≥99%
Certificats
4-Fluorophenol ≥99%

3 Options disponibles ci-dessous

Product Details & Documents

Purity: 99% CAS No.: 371-41-5 Molecular Formula: C6H5FO Formula Weight: 112.10 Flash Point: 68°(154°F) Odour: Phenol-like Boiling Point: 185-188° Beilstein Reference: 1362752 UN No.: 2923 MDL No.: MFCD00002316

Synonymes:  p-Fluorophenol, 1-Fluoro-4-hydroxybenzene
  • Numéro CAS:371-41-5
  • Formule:C₆H₅FO
  • Numéro MDL:MFCD00002316
  • Poids moléculaire:112.1 g/mol
  • Point d'ébullition:185…188 °C
  • Point de fusion:44…50 °C
  • Densité:1.22
  • Point d'éclair:68 °C (154 °F)
  • Température de stockage:Ambient
  • EINECS:206-736-0
  • UN:2923
  • ADR:8,III
Product Family Options

Informations sur le produit

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.