Orders LinkContactUsLinkComponent
Weighing dishes, anti-static
Supplier:   Simport Scientific
Weighing dishes, anti-static
Supplier:  Simport Scientific
Certificates
Weighing dishes, anti-static
Weighing dishes, anti-static
3 Options Available Below
Product Details & Documents
These weighing dishes are safe, contaminant-free, biologically inert economical containers for weighing liquid or powdered samples in the laboratory.
  • Resistant to diluted acids, aqueous solutions, alcohols and bases
  • Flat bottom ensures perfect stability on countertops
  • Smooth surface providing accurate pour-outs with minimal sample loss
  • Facilitate weighing of static-affected samples
  • Ideal for many applications such as weighing, dispensing or storing
Dishes can also be used as quick freeze trays for sample material, discard trays for broken ampules, or mixing trays for small batches. Withstand temperatures up to 80 °C.
Product Family Options
Product Information
  • Capacity
    W×D×H
    Colour
    Availability
    Price
  • 10 ml
    40×40×8 mm
    White
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 26,60 €
    26,60 €
    1 * 500 ST
       26,60 €
  • 100 ml
    78×78×25 mm
    White
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 73,80 €
    73,80 €
    1 * 500 ST
       73,80 €
  • 300 ml
    127×127×25 mm
    White
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 152,00 €
    152,00 €
    1 * 500 ST
       152,00 €