Micro spoon
With plastic handle, 150 mm, 5 mm
RSGA087.025
: RSG
This micro spoon is made of stainless steel (Remanit 4301) and features a black plastic handle.
- Description:With plastic handle
- Length:150 mm
- Spoon size:5 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



