Orders LinkContactUsLinkComponent
Cooling Boxes
Cooling Boxes
Supplier:   KARTELL
Cooling Boxes
Supplier:  KARTELL
Certificates
Cooling Boxes
2 Options Available Below
Product Details & Documents
Cooling boxes are low temperature containers made of high impact polystyrene material, thermally isolated by means of a polyurethane isolating jacket.
  • Non-toxic
  • 100% food safe
Delivery information: Cooling block is available for cooling boxes.
Product Family Options
Product Information
  • Capacity
    Colour
    Dimensions
    Availability
    Price
  • 25 l
    Dark anthracite
    230×400×390 mm (ext); 180×350×375 mm (int)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 190,00 €
    190,00 €
    1 * 1 ST
       190,00 €
  • 32 l
    280×450×380 mm (ext); 230×400×320 mm (int)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 230,00 €
    230,00 €
    1 * 1 ST
       230,00 €
Accessories
Product Information
  • Catalog #
    Capacity
    W×D×H
    Availability
    Price
  • HDPE cooling block for Kartell Frigo box
    HDPE cooling block for Kartell Frigo box
    KART3707
    500 ml
    17,5×9×3 cm
    Log in for availability
    1 * 1 ST - 10,90 €