Orders LinkContactUsLinkComponent
Sample cups, SeraNest™
Sample cups, SeraNest™
Fournisseur:   Simport Scientific
Sample cups, SeraNest™
Fournisseur:  Simport Scientific
Certificates
Sample cups, SeraNest™
Sample cups, SeraNest™
2 Options Available Below
Product Details & Documents
PS, clear.
  • Good chemical resistance
  • Good temperature resistance
These sample cups will hold perfectly on top of blood collection tubes. Just pour the blood sample into a SeraNest™ and its low shoulder will hold securely on the tube. No need to relabel. Two sizes available.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Product Family Options
Product Information
  • Capacity
    Ext.Ø
    Packed
    Disponibilité
    Prix
  • SIMPB720-13
    For blood collection tubes
    For blood collection tubes
    1,0 ml
    13,9 mm
    Bag of 1000, Carton of 5000
    Log in for availability
    Log in for availability
    1 * 5.000 ST
    1 * 5.000 ST - 379,00 €
    379,00 €
    1 * 5.000 ST
       379,00 €
  • SIMPB720-16
    For blood collection tubes
    For blood collection tubes
    2,0 ml
    16,7 mm
    Bag of 1000, Carton of 5000
    Log in for availability
    Log in for availability
    1 * 5.000 ST
    1 * 5.000 ST - 558,00 €
    558,00 €
    1 * 5.000 ST
       558,00 €