Orders LinkContactUsLinkComponent
Eyewash bottle
Eyewash bottle
Empty eyewash bottle, PE, 500 ml, with specially designed snap-on cap
Catalog # 215-1363
Supplier:   KARTELL
Eyewash bottle
Catalog # 215-1363
Supplier:  KARTELL
Eyewash bottle
Eyewash bottle
Empty eyewash bottle, PE, 500 ml, with specially designed snap-on cap
Price
26,90 €
Log in for availability
1 * 1 ST
Product Details & Documents
This eyewash bottle can be filled with distilled water or other medicated liquid for ophthalmic use and allows a quick and immediate flushing.
  • Provided with an eye cup with snap on cap
  • Airvent valve in the head assures air refill, thus preventing draw back of contaminated wash into the bottle
  • perforated spray plate in the cup centre breaks up the stream to give the eye a gentle wash
  • Drain tube releases waste
Specifications
  • Description:
    Empty eyewash bottle, PE, 500 ml, with specially designed snap-on cap
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.