Order Entry
Belgium
ContactUsLinkComponent
Peripheral IV cannulae, Insyte™ Autoguard™
Peripheral IV cannulae, Insyte™ Autoguard™
  BDAM381954
Peripheral IV cannulae, Insyte™ Autoguard™
  BDAM381954

 

  • Ø×L:
    1,7× 30 mm
  • Gauge:
    G 16
  • Colour:
    Grey
  • Description:
    Safety peripheral IV cannulae
  • Seq:
    01
  • Pk:
    200

 

 

Sterile disposable cannulae made of bio materials.

  • Automatic stop of the blood
  • Safety technology reducing by 95% the risks of injuries
  • Apparels are protected and cross contamination is avoided

Suitable for blood controls.