Orders LinkContactUsLinkComponent
Buckets, Round, HDPE
Supplier:   BURKLE
Buckets, Round, HDPE
Supplier:  BURKLE
Certificates
Buckets, Round, HDPE
Buckets, Round, HDPE
4 Options Available Below
Product Details & Documents
HDPE, white or black.
  • With litre scale, graduation 1000 ml
  • With spout
  • White buckets suitable for use with foodstuffs
  • Black buckets not suitable for food
For transporting, decanting and mixing. Wide spout gives rapid but safe emptying. With plastic handle.
Product Family Options
Product Information
  • Division
    Colour
    Capacity
    Ø
    Height
    Availability
    Price
  • White
    10,5 L
    286 mm
    289 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 41,20 €
    41,20 €
    1 * 1 ST
       41,20 €
  • Black
    10,5 L
    286 mm
    289 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 36,50 €
    36,50 €
    1 * 1 ST
       36,50 €
  • White
    17 L
    330 mm
    315 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 48,90 €
    48,90 €
    1 * 1 ST
       48,90 €
  • 1000 ml
    Black
    17 L
    330 mm
    315 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 43,60 €
    43,60 €
    1 * 1 ST
       43,60 €