Orders LinkContactUsLinkComponent
pH combination electrodes, ProcessLine
pH combination electrodes, ProcessLine
Supplier:   SI Analytics
pH combination electrodes, ProcessLine
Supplier:  SI Analytics
Certificates
pH combination electrodes, ProcessLine
pH combination electrodes, ProcessLine
41 Options Available Below
Product Details & Documents
These electrodes are low maintenance sensors for heavy duty process applications in the chemical industry. They are ideally suitable for measuring media with extreme ionic strength - whether boiler feed water or brine - also in strongly oxidizing acid and alkali containing media.
  • Low-maintenance electrodes: no electrolyte topping up or installation of complicated pressure sequence control required
  • Pinhole diaphragms, so reference electrode is not polluted or blocked
  • Duralid electrolyte with high KCl content combines very long working life with rapid and stable measurements
  • Proven H membrane glass with very low alkali error and optimized spherical shape
  • Integrated pressure compensation cushion
PL8× models
Reference system: Duralid solid electrolyte with a high KCl content
Diaphragm: 2-hole
PL9× models
Reference system: RheoLid electrolyte with KCI storage rings
Diaphragm: PTFE
Product Family Options
Product Information
  • Type
    pH
    Temp.
    Material
    Ø×L
    Connection
    Availability
    Price
  • PL 80-120pH
    0 - 14
    0...+130 °C
    Glass
    12×120 mm
    S8
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 341,00 €
    341,00 €
    1 * 1 ST
       341,00 €
  • PL 80-225pH
    0 - 14
    0...+130 °C
    Glass
    12×225 mm
    S8
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 420,00 €
    420,00 €
    1 * 1 ST
       420,00 €
  • PL 80-325pH
    0 - 14
    0...+130 °C
    Glass
    12×325 mm
    S8
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 456,00 €
    456,00 €
    1 * 1 ST
       456,00 €
  • PL 80-425pH
    0 - 14
    0...+130 °C
    Glass
    12×225 mm
    S8
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 506,00 €
    506,00 €
    1 * 1 ST
       506,00 €
  • PL 81-120pHT VP, with Pt1000
    0 - 14
    0...+130 °C
    Glass
    12×120 mm
    VP screw plug
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 424,00 €
    424,00 €
    1 * 1 ST
       424,00 €
  • PL 81-225pHT VP, with Pt1000
    0 - 14
    0...+130 °C
    Glass
    12×225 mm
    VP screw plug
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 504,00 €
    504,00 €
    1 * 1 ST
       504,00 €
  • PL 81-325pHT VP, with Pt1000
    0 - 14
    0...+130 °C
    Glass
    12×325 mm
    VP screw plug
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 540,00 €
    540,00 €
    1 * 1 ST
       540,00 €
  • PL 81-360pHT VP, with Pt1000
    0 - 14
    0...+130 °C
    Glass
    12×360 mm
    VP screw plug
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 573,00 €
    573,00 €
    1 * 1 ST
       573,00 €
  • PL 81-425pHT VP, with Pt1000
    0 - 14
    0...+130 °C
    Glass
    12×425 mm
    VP screw plug
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 589,00 €
    589,00 €
    1 * 1 ST
       589,00 €
  • PL 82-120pHT VP, with Pt100
    0 - 14
    0...+130 °C
    Glass
    12×120 mm
    VP screw plug
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 456,00 €
    456,00 €
    1 * 1 ST
       456,00 €
  • PL 82-225pHT VP, with Pt100
    0 - 14
    0...+130 °C
    Glass
    12×225 mm
    VP screw plug
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 508,00 €
    508,00 €
    1 * 1 ST
       508,00 €
  • PL 82-325pHT VP, with Pt100
    0 - 14
    0...+130 °C
    Glass
    12×325 mm
    VP screw plug
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 550,00 €
    550,00 €
    1 * 1 ST
       550,00 €
  • PL 82-360pHT VP, with Pt100
    0 - 14
    0...+130 °C
    Glass
    12×360 mm
    VP screw plug
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 585,00 €
    585,00 €
    1 * 1 ST
       585,00 €
  • PL 82-425pHT VP, with Pt100
    0 - 14
    0...+130 °C
    Glass
    12×425 mm
    VP screw plug
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 594,00 €
    594,00 €
    1 * 1 ST
       594,00 €
  • PL 83-225 NMSN, with NTC30
    0 - 14
    0...+130 °C
    Glass
    12×225 mm
    Memosens® screw plug
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 596,00 €
    596,00 €
    1 * 1 ST
       596,00 €
  • PL 83-325 NMSN, with NTC30
    0 - 14
    0...+130 °C
    Glass
    12×325 mm
    Memosens® screw plug
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 636,00 €
    636,00 €
    1 * 1 ST
       636,00 €
  • PL 83-360 NMSN, with NTC30
    0 - 14
    0...+130 °C
    Glass
    12×360 mm
    Memosens® screw plug
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 661,00 €
    661,00 €
    1 * 1 ST
       661,00 €
  • PL 83-425 NMSN, with NTC30
    0 - 14
    0...+130 °C
    Glass
    12×425 mm
    Memosens® screw plug
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 680,00 €
    680,00 €
    1 * 1 ST
       680,00 €
  • PL 89-120Pt, redox
    0 - 14
    0...+130 °C
    Glass
    12×120 mm
    Pg 13.5
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 504,00 €
    504,00 €
    1 * 1 ST
       504,00 €
  • PL 89-225Pt, redox
    0 - 14
    0...+130 °C
    Glass
    12×225 mm
    S8
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 551,00 €
    551,00 €
    1 * 1 ST
       551,00 €
  • PL 89-325Pt, redox
    0 - 14
    0...+130 °C
    Glass
    12×325 mm
    S8
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 594,00 €
    594,00 €
    1 * 1 ST
       594,00 €
  • PL 89-120 NMSN, with NTC30
    0...+130 °C
    Glass
    12×120 mm
    Memosens® screw plug
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 604,00 €
    604,00 €
    1 * 1 ST
       604,00 €
  • PL 89-225 NMSN, with NTC30
    0...+130 °C
    Glass
    12×225 mm
    Memosens® screw plug
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 654,00 €
    654,00 €
    1 * 1 ST
       654,00 €
  • PL-S 82-225pHT VP, with Pt100
    0 - 14
    0...+130 °C
    Glass
    12×225 mm
    VP screw plug
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 569,00 €
    569,00 €
    1 * 1 ST
       569,00 €
  • PL 99-120 Pt
    0 - 14
    –5...135 °C
    Glass
    12×120 mm
    S8
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 511,00 €
    511,00 €
    1 * 1 ST
       511,00 €
  • PL 99-225 Pt
    0 - 14
    –5...135 °C
    Glass
    12×225 mm
    S8
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 572,00 €
    572,00 €
    1 * 1 ST
       572,00 €
  • PL 99-120 NMSN, with NTC30
    0 - 14
    –5...135 °C
    Glass
    12×120 mm
    Memosens® screw plug
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 696,00 €
    696,00 €
    1 * 1 ST
       696,00 €
  • PL A-90-120 pH
    0 - 14
    0...+110 °C
    Glass
    12×120 mm
    S8
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 378,00 €
    378,00 €
    1 * 1 ST
       378,00 €
  • PL A-91-120 pHT VP, with Pt1000
    0 - 14
    –5...+110 °C
    Glass
    12×120 mm
    VP screw plug
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 456,00 €
    456,00 €
    1 * 1 ST
       456,00 €
  • PL A-91-225 pHT VP, with Pt1000
    0 - 14
    –5...+110 °C
    Glass
    12×225 mm
    VP screw plug
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 507,00 €
    507,00 €
    1 * 1 ST
       507,00 €
  • PL A-93-120 NMSN, with NTC30
    0 - 14
    –5...+110 °C
    Glass
    12×120 mm
    Memosens® screw plug
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 530,00 €
    530,00 €
    1 * 1 ST
       530,00 €
  • PL H-90-120 pH
    0 - 14
    0...+135 °C
    Glass
    12×120 mm
    S8
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 387,00 €
    387,00 €
    1 * 1 ST
       387,00 €
  • PL H-90-225 pH
    0 - 14
    0...+135 °C
    Glass
    12×225 mm
    S8
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 445,00 €
    445,00 €
    1 * 1 ST
       445,00 €
  • PL H-90-360 pH
    0 - 14
    0...+135 °C
    Glass
    12×360 mm
    S8
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 517,00 €
    517,00 €
    1 * 1 ST
       517,00 €
  • PL H-91-120 pHT VP, with Pt1000
    0 - 14
    0...+135 °C
    Glass
    12×120 mm
    VP screw plug
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 463,00 €
    463,00 €
    1 * 1 ST
       463,00 €
  • PL H-91-225 pHT VP, with Pt1000
    0 - 14
    0...+135 °C
    Glass
    12×225 mm
    VP screw plug
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 517,00 €
    517,00 €
    1 * 1 ST
       517,00 €
  • PL H-93-120 NMSN, with NTC30
    0 - 14
    0...+135 °C
    Glass
    12×120 mm
    Memosens® screw plug
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 553,00 €
    553,00 €
    1 * 1 ST
       553,00 €
  • PL H-93-225 NMSN, with NTC30
    0 - 14
    0...+135 °C
    Glass
    12×225 mm
    Memosens® screw plug
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 625,00 €
    625,00 €
    1 * 1 ST
       625,00 €
  • PL H-93-325 NMSN, with NTC30
    0 - 14
    0...+135 °C
    Glass
    12×325 mm
    Memosens® screw plug
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 659,00 €
    659,00 €
    1 * 1 ST
       659,00 €
  • PL H-93-360 NMSN, with NTC30
    0 - 14
    0...+135 °C
    Glass
    12×360 mm
    Memosens® screw plug
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 696,00 €
    696,00 €
    1 * 1 ST
       696,00 €
  • PL PETR-120VP, with Pt1000 + redox
    0 - 14
    0...+130 °C
    Glass
    12×120 mm
    VP screw plug
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 586,00 €
    586,00 €
    1 * 1 ST
       586,00 €